Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-586

Sigma-Aldrich

MAPKAP Kinase 3 Protein, unactive, 50 µg

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$685.00

$685.00


Please contact Customer Service for Availability


Select a Size

Change View
50 μG
$685.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

$685.00


Please contact Customer Service for Availability

biological source

human

Quality Level

mol wt

Mw ~69.8 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

General description

N-terminal, GST-tagged, recombinant, human MAPKAP Kinase 3, amino acids 2-end
expressed in E.coli cells

Application

Activation Assay
Research Category
Metabolism
Research Sub Category
Obesity|Metabolic Disorders
Unactive, N-terminal, GST-tagged, recombinant, human MAPKAP Kinase 3, amino acids 2-end, for use in Kinase Assays.

Biochem/physiol Actions

  • Protein Target: MAPKAP-K3
  • Target Sub-Family:CAMK

Quality

Routinely evaluated to activate SAPK2a (14-587).

Physical form

Format: Glutathione agarose affinity chromatography

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service