Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-480-M

Sigma-Aldrich

Rsk2/MAPKAP Kinase 1b Protein, active, 10 µg

Active, recombinant human RSK2, full-length with an N-terminal His6 tag between Met1 & Pro2, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

biological source

human

Quality Level

100
300

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 88 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... RPS6KA3(6197)

General description

Recombinant human RSK2, full-length with an N-terminal His6 tag between Met1 and Pro2

Packaging

Also available in 250 µg size --call for pricing and availability and reference catalog number 14-480M when ordering the 250 µg size.

Quality

the MAPKAP Kinase 2 substrate peptide

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yiguo Zhang et al.
The Journal of biological chemistry, 278(15), 12650-12659 (2003-02-04)
Phosphorylation at Ser(727) is known to be required for complete activation of STAT3 by diverse stimuli including UV irradiation, but the kinase(s) responsible for phosphorylating STAT3 (Ser(727)) is still not well discerned. In the present study, we observed that activation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service