Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.44235

Millipore

Nozzle Set

for Compressed gas testing with RCS® High Flow Touch, for use with RCS® High Flow Touch

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NA.74

material

stainless steel nozzle

Agency

non-compendial

product line

RCS®

feature

adjustable

packaging

pkg of 1 kit

application(s)

compressed gas monitoring
cosmetics
food and beverages
pharmaceutical

shipped in

ambient

storage temp.

room temp

General description

Nozzle Set is an additional accessory for use with RCS® Compressed Gas Adapter. The RCS® High Flow Touch is a portable air sampler for fast and easy monitoring of ambient air and compressed gases in controlled environments. The microbial air sampler is based on impaction by centrifugal force. A Set of five nozzles are provided to extend the air inlet pressure from 1 bar to 0.1-7.0 bar.To be combined with the RCS Compressed Gas Adapter for microbial monitoring of compressed gasses.

Application

Nozzle Set is an additional accessory with the RCS® microbial air sampler used to extend the air inlet pressure from 1 bar to 0.1-7.0 bar. These air samplers find application in microbial monitoring of ambient air in pharmaceutical, medical and food industry.

Compatibility

The Nozzle Set is compatible for use with RCS® Air Samplers of the High Flow series (RCS® High Flow Touch, RCS® High Flow) and RCS® Isolator.

Legal Information

RCS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Ensure microbial air sampling generates precise results complying with international standards. Our systems are easy-to-use and reliable.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service