Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-330

Sigma-Aldrich

JNK2α2/SAPK1a Protein, inactive, 50 g

Unactive, N-terminal His6-tagged, recombinant, full-length, human JNK2α2/SAPK1a unactive, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$546.00

$546.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
$546.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

$546.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in Sf21 cells

mol wt

Mw 49.2 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

UniProt accession no.

shipped in

dry ice

Gene Information

human ... MAPK9(5601)

General description

N-terminal His6-tagged, recombinant, full-length, human JNK2α2/SAPK1a unactive
Product Source: Expressed in Sf21 insect cells

Biochem/physiol Actions

Protein Target: JNK2α2/SAPK1a
Target Sub-Family: CMGC

Quality

Routinely evaluated by phosphorylation of JNK2α2/SAPK1a

Physical form

Ni2+/NTA-agarose

Storage and Stability

1 year at -20°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Gupta et al.
Science (New York, N.Y.), 267(5196), 389-393 (1995-01-20)
Treatment of cells with pro-inflammatory cytokines or ultraviolet radiation causes activation of the c-Jun NH2-terminal protein kinase (JNK). Activating transcription factor-2 (ATF2) was found to be a target of the JNK signal transduction pathway. ATF2 was phosphorylated by JNK on
S Gupta et al.
The EMBO journal, 15(11), 2760-2770 (1996-06-03)
The JNK protein kinase is a member of the MAP kinase group that is activated in response to dual phosphorylation on threonine and tyrosine. Ten JNK isoforms were identified in human brain by molecular cloning. These protein kinases correspond to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service