Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.43023

Supelco

Sulfuric acid solution

0.02 N

Sign Into View Organizational & Contract Pricing


About This Item

form

liquid

concentration

0.00999-0.01001 (mol/l)

color

≤10, APHA

suitability

suitable for odor (odorless)

General description

Volumetric solution

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service