Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-246-M

Sigma-Aldrich

p38γ/SAPK3 Protein, active, 10 µg

Active, recombinant GST fusion protein corresponding to full length human p38γ/SAPK3, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
$5,190.00

$5,190.00


Please contact Customer Service for Availability


Select a Size

Change View
250 μG
$5,190.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

$5,190.00


Please contact Customer Service for Availability

biological source

human

Quality Level

recombinant

expressed in E. coli

mol wt

Mw 69 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... PEBP1(5037)

General description

Product Source: expressed in E. coli
Recombinant GST fusion protein corresponding to full length human p38γ/SAPK3

Biochem/physiol Actions

Protein Target: p38γ/SAPK3
Target Sub-Family: CMGC

Quality

Routinely evaluated by phosphorylation of MBP in vitro

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The hepatitis B virus HBx protein inhibits caspase 3 activity
Gottlob, K., et al
The Journal of Biological Chemistry, 273, 33347-33353 (1998)
A Cuenda et al.
The EMBO journal, 16(2), 295-305 (1997-01-15)
Stress-activated protein kinase-3 (SAPK3), a recently described MAP kinase family member with a wide-spread tissue distribution, was transfected into several mammalian cell lines and shown to be activated in response to cellular stresses, interleukin-1 (IL-1) and tumour necrosis factor (TNF)
S P Davies et al.
The Biochemical journal, 351(Pt 1), 95-105 (2000-09-22)
The specificities of 28 commercially available compounds reported to be relatively selective inhibitors of particular serine/threonine-specific protein kinases have been examined against a large panel of protein kinases. The compounds KT 5720, Rottlerin and quercetin were found to inhibit many
Y Fleming et al.
The Biochemical journal, 352 Pt 1, 145-154 (2000-11-04)
Stress-activated protein kinase 1 (SAPK1), also called c-Jun N-terminal kinase (JNK), becomes activated in vivo in response to pro-inflammatory cytokines or cellular stresses. Its full activation requires the phosphorylation of a threonine and a tyrosine residue in a Thr-Pro-Tyr motif

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service