Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-1048

Sigma-Aldrich

Human recombinant CARM1

Human recombinant CARM1 with an N-terminal FLAG tag expressed in FreeStyle 293-F cells.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$651.00

$651.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$651.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

$651.00


Please contact Customer Service for Availability

Request a Bulk Order

recombinant

expressed in FreeStyle 293-F cells

Quality Level

mol wt

Mw ~66 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (histone methyltransferase (HMT))

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... CARM1(10498)

General description

Product Source: Human

Application

Human recombinant CARM1 with an N-terminal FLAG tag expressed in FreeStyle 293-F cells.
Research Category
Nuclear Function

Quality

routinely evaluated by methylation of histone H3

Linkage

Replaces: 14-575

Storage and Stability

6 months at -80°C.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.
Varies by lot; see lot-specific datasheet for information on each lot.

Legal Information

FreeStyle is a trademark of Invitrogen Corp.
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongwei Li et al.
The Journal of biological chemistry, 277(47), 44623-44630 (2002-09-19)
The RNA-binding protein HuR stabilizes labile mRNAs carrying AU-rich instability elements. This mRNA stabilization can be induced by hypoxia, lipopolysaccharide, and UV light. The mechanism by which these stimuli activate HuR is unclear and might be related to post-translational modification
Melody D Fulton et al.
The Journal of biological chemistry, 297(4), 101123-101123 (2021-09-08)
Histone arginine methylation is a key post-translational modification that mediates epigenetic events that activate or repress gene transcription. Protein arginine methyltransferases (PRMTs) are the driving force for the process of arginine methylation, and the core histone proteins have been shown
Seth Frietze et al.
Cancer research, 68(1), 301-306 (2008-01-04)
Estrogen receptor alpha (ER alpha) mediates breast cancer proliferation through transcriptional mechanisms involving the recruitment of specific coregulator complexes to the promoters of cell cycle genes. The coactivator-associated arginine methyltransferase CARM1 is a positive regulator of ER alpha-mediated transcriptional activation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service