Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

138001

Sigma-Aldrich

D-Amino Acid Oxidase Inhibitor II, CBIO

The D-Amino Acid Oxidase Inhibitor II, CBIO, also referenced under CAS 61977-29-5, controls the biological activity of D-Amino Acid Oxidase.

Synonym(s):

D-Amino Acid Oxidase Inhibitor II, CBIO, 6-Chloro-1,2-benzisoxazol-3(2H)-one, 6-Chlorobenzo[d]isoxazol-3-ol, D-AAO Inhibitor II, CBIO

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$126.00

$126.00


Please contact Customer Service for Availability


Select a Size

Change View
50 MG
$126.00

About This Item

Empirical Formula (Hill Notation):
C7H4ClNO2
CAS Number:
Molecular Weight:
169.57
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$126.00


Please contact Customer Service for Availability

Quality Level

Assay

≥97% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Clc1cc2[o][nH][c](c2cc1)=O

InChI

1S/C7H4ClNO2/c8-4-1-2-5-6(3-4)11-9-7(5)10/h1-3H,(H,9,10)

InChI key

SJAPSPJRTQCDNO-UHFFFAOYSA-N

General description

A cell-permeable benzisoxazolone compound that potently inhibits D-amino acid oxidase (DAO/DAAO/DAMOX/OXDA) activity (IC50 = 90 and 150/188 nM against rat spinal cord and porcine kidney DAO, respectively) in a reversible and D-serine-competitive (Ki = 100 nM) manner. Upon co-administration, CBIO (30 mg/kg, p.o.) markedly prolongs the half-life of orally administered D-serine (30 mg/kg) in plasma, resulting in enhanced rat brain D-serine level (60% vs. 25% above basal, respectively, with vs. without CBIO). Direct intrathecal/spinal CBIO injection either before (ED50 =60 ng) or after (ED50 = 170 ng) subcutaneous formalin injection (50 µl 5% solution) greatly alleviates formalin-induced late phase tonic pain (Emax =67%), but not early phase acute nociception in rats in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
It is recommended to prepare a stock solution for 6 month use each time and store the unreconstituted material in solid form, protected from moisture and preferably under inert gas, for best long-term stability during storage.

Other Notes

Gong, N., et al. 2011. J. Pharmacol. Exp. Ther.336, 282.
Ferraris, D., et al. 2008. J. Med. Chem.51, 3357.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service