Skip to Content
MilliporeSigma

137133

SAFC

Ethanolamine

EMPROVE® EXPERT, NF

Pharma Manufacturing

Synonym(s):

2-Aminoethanol, 2-Aminoethyl alcohol, ETA, MEA, MEA 90, MEA-LCI, Monoethanolamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$538.00
20 L
$5,801.00

$538.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 L
$538.00
20 L
$5,801.00

About This Item

Linear Formula:
NH2CH2CH2OH
CAS Number:
Molecular Weight:
61.08
Beilstein:
505944
MDL number:
UNSPSC Code:
12352116

$538.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest a Sample

biological source

synthetic

Quality Level

Agency

NF

vapor density

2.1 (vs air)

vapor pressure

0.2 mmHg ( 20 °C)

description

Product will be discontinued and replaced by 1.37044

product line

EMPROVE® EXPERT

form

liquid

autoignition temp.

1436 °F

expl. lim.

17 %

packaging

pkg of 1 L (1371331000)
pkg of 20 L (1371339020)

refractive index

n20/D 1.454 (lit.)

bp

170 °C (lit.)
69-70 °C/10 mmHg

mp

10-11 °C (lit.)

density

1.012 g/mL at 25 °C (lit.)

storage temp.

2-25°C

SMILES string

NCCO

InChI

1S/C2H7NO/c3-1-2-4/h4H,1-3H2

InChI key

HZAXFHJVJLSVMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

195.8 °F - Pensky-Martens closed cup

Flash Point(C)

91 °C - Pensky-Martens closed cup


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service