Skip to Content
MilliporeSigma
All Photos(5)

Documents

1.32360

Millipore

MC-Media Pad®

for yeasts, for molds, Accredited for Food Stuff and Dried Cannabis Flower (THC>0.3%)

Synonym(s):

Yeast and Mold count test

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171504
NACRES:
NA.75

Agency

AOAC® OMA
AOAC® PTM
MicroVal

sterility

sterile

product line

MC-Media Pad®

feature

ready-to-use

pH

7.4 ( in H2O)

application(s)

food and beverages
personnel monitoring
surface monitoring
water monitoring

shipped in

ambient

storage temp.

2-25°C

suitability

molds
yeasts

General description

The MC MEDIA PAD® offers a convenient solution for rapidly and routinely testing microbial contamination in raw and in-process food and beverage materials, as well as finished products. The MC MEDIA PAD® is a pre-sterilized, ready-to-use dry culture device, simplifying testing and minimizing the quantity of waste. MC MEDIA PAD® is composed of a unique adhesive sheet, a test pad coated with medium and water absorption polymer, and a transparent cover film, to automatically re-constitute the media. MC MEDIA PAD® Yeast & Mold is intended for enumeration of yeasts and molds in foods with an aw of >0.95. The MC MEDIA PAD® is coated with a growth medium and a redox indicator for detection. Once the liquid sample is inoculated onto the test pad, the sample diffuses through the whole pad by capillary action.

Application

MC-Media Pad® Yeasts and Molds is recommended for the detection and enumeration of Yeasts and molds in foods, beverages, and dried Cannabis Flower (THC>0.3%).

Features and Benefits

  • Compliant with international food & beverage regulations
  • Reduce waste and save space
  • Reliable and fast results
  • Easy to use—fewer handling steps than traditional methods
  • Colour coded to ensure the use of right Media Pad
  • Simplified workflow

Analysis Note

If target organisms are present, they grow as red to reddish brown coloured colonies on the test pad.

Legal Information

AOAC is a registered trademark of AOAC International
MC MEDIA PAD is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This article will focus on results obtained for MC-Media Pad RAC, MC-Media Pad Coliform, MC-Media Pad E. coli & Coliform & MC-Media Pad Yeast & Mold.

Protocols

Comparison of MC-Media Pad® with different convenience solution products for indicator organism testing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service