Skip to Content
MilliporeSigma

1.28225

SAFC

Sodium sulfate anhydrous

EMPROVE® EXPERT, Ph. Eur., BP, ChP, USP

Pharma Manufacturing

Synonym(s):

Sodium sulfate

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$282.00
3 X 1 KG
$562.00

$282.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 KG
$282.00
3 X 1 KG
$562.00

About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
MDL number:
UNSPSC Code:
51171629
EC Index Number:
231-820-9

$282.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest a Sample

grade

anhydrous

Quality Level

Agency

BP
ChP
Ph. Eur.
USP

product line

EMPROVE® EXPERT

form

solid

mol wt

142.04 g/mol

impurities

endotoxins, low

pH

5.2-8.0 (20 °C, 50 g/L in H2O)

bp

884 °C (lit.)

mp

884 °C (lit.)
884 °C

solubility

200 g/L

density

2.68 g/cm3 at 25 °C
2.68 g/mL at 25 °C (lit.)

application(s)

pharma/biopharma processes
pharmaceutical

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Sodium sulfate anhydrous is the anhydrous, sodium salt form of sulfuric acid, and can be used as buffer and pH modifier or mineral salt.
Due to its low microbial and endotoxin limits, Sodium sulfate anhydrous Emprove® Expert is especially suitable for high-risk applications.

Packaging

1282250003//3X1kg - plastic bottle
1282251001//1kg - plastic bottle
1282258012//12kg - plastic bottle
1282258026//25kg - plastic drum

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service