Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12-570

Sigma-Aldrich

Monomethyl-Histone H3 Peptide

Lys36, biotin conjugate

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$410.00

$410.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
$410.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.42

$410.00


Please contact Customer Service for Availability

Product Name

Monomethyl-Histone H3 (Lys36) Peptide, biotin conjugate, Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes.

biological source

synthetic

Quality Level

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... H3F3B(3021)

Application

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes.

Storage and Stability

stable 2 years at -20°C from date of shipment

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The ciliary smooth muscle electrotransfer: basic principles and potential for sustained intraocular production of therapeutic proteins.
Elodie Touchard,Laura Kowalczuk,Carole Bloquel,Marie-Christine Naud,Pascal Bigey et al.
The journal of gene medicine null
Expression of stress proteins and lymphocyte reactivity in heterotopic cardiac allografts undergoing cellular rejection.
J Qian,R Moliterno,M A Donovan-Peluso,K Liu,J Suzow,L Valdivia,F Pan,R J Duquesnoy
Transplant Immunology null
Hanqing Lin et al.
Cell research, 20(8), 899-907 (2010-06-23)
H3K9me2 and H3K27me2 are important epigenetic marks associated with transcription repression, while H3K4me3 is associated with transcription activation. It has been shown that active and repressive histone methylations distribute in a mutually exclusive manner, but the underlying mechanism was poorly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service