Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12-566

Sigma-Aldrich

Dimethyl-Histone H3 Peptide

Lys27, biotin conjugate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.42

product name

Dimethyl-Histone H3 (Lys27) Peptide, biotin conjugate, Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes.

Quality Level

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... H3F3B(3021)

Storage and Stability

stable 2 years at -20°C from date of shipment

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service