Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12-540

Sigma-Aldrich

LKBtide

LKBtide primarily used in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$390.00

$390.00


Please contact Customer Service for Availability


Select a Size

Change View
1 MG
$390.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.42

$390.00


Please contact Customer Service for Availability

Quality Level

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

Description: Synthetic peptide (LSNLYHQGKFLQTFCGSPLYRRR) corresponding to amino aicds 196-215 of human NUAK2, plus 3 additional arginine residues.

MW: 2783 Daltons.

Biochem/physiol Actions

Protein Target: LKB1
Target Sub-Family: CAMK

Quality

Routinely evaluated as a substrate for LKB1/STRADa/MO25a, active (Catalog # 14-596).

Physical form

Lyophilized powder

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service