Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

125188M

Millipore

Eshmuno® CMX Resin

RoboColumn Eshmuno® CMX, volume 0.6 mL, prepacked column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$494.00

$494.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$494.00

About This Item

UNSPSC Code:
41115711

$494.00


Please contact Customer Service for Availability

ligand

(Carboxyethyl with Alkyl-Functional Groups)

material

polypropylene housing

sterility

non-sterile (
sterile)

product line

Eshmuno®

form

prepacked column

parameter

≤8 bar pressure
300 cm/hr max. flow rate

volume

0.6 mL

matrix active group

Polyvinyl Ether

mean particle size

50 μm

capacity

~60 mL, resin binding capacity

application(s)

ADC
mAb
recombinant protein

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

Eshmuno® CMX chromatography resin is a mixed mode chromatography resin built on the proven Eshmuno® resin technology. This innovative resin combines weak cation exchange properties with hydrophobic interaction, providing high selectivity for Monoclonal Antibody (mAb), fusion protein and Antibody Drug Conjugate (ADCs) purification as well as separation of low molecular weight impurities, Host Cell Proteins (HCPs) and glycovariants.

Application

Purification of mAbs, ADCs, fusion proteins

Features and Benefits

Eshmuno® CMX chromatography resin enables users to
  • Intensify processes and decrease costs by reducing the number of chromatographic steps and buffer consumption
  • Improve performance with higher recovery rates, high selectivity and superior dynamic binding capacity
  • Improve the user experience with a broad operational window, simplified process development and easy column packing due to a rigid base bead

Legal Information

Eshmuno is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Learn more on mAb downstream processing, more specifically polishing chromatography and the relevant associated products.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service