Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.25150

Millipore

Fractogel® EMD TMAE Medcap (M)

RoboColumn Fractogel® TMAE MC (M), volume 0.6 mL, prepacked column

Synonym(s):

RoboCol Fractogel® TMAE MC (M) 0.6ml

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$494.00

$494.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$494.00

About This Item

UNSPSC Code:
41115711

$494.00


Please contact Customer Service for Availability

ligand

(Trimethylammoniumethyl)

Quality Level

material

polypropylene housing

product line

Fractogel®

form

prepacked column

parameter

≤8 bar pressure
~180 cm/hr max. flow rate

column I.D. × L

5 mm × 30 mm

volume

0.6 mL

matrix active group

Methacrylate

mean particle size

40-90 μm

capacity

~150 mg, BSA binding capacity

application(s)

plasma purification
recombinant protein
vaccine development

separation technique

anion exchange

storage temp.

no temp limit

Looking for similar products? Visit Product Comparison Guide

General description

5 x 30 mm

Features and Benefits

RoboCol Fractogel® TMAE MedCap (M) 0.6 ml enables:
  • Excellent binding at high flow rates
  • Excellent pressure-flow behavior
  • Compatibility with 2.5 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl storage solution

Legal Information

FRACTOGEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service