Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.25149

Millipore

Fractogel® EMD Chelate (M)

RoboColumn Fractogel® Chelate (M), volume 0.2 mL, prepacked column

Synonym(s):

RoboColumn Fractogel® Chelate (M) 0.2ml

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$418.00

$418.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$418.00

About This Item

UNSPSC Code:
41115711

$418.00


Please contact Customer Service for Availability

ligand

iminodiacetic acid

material

polypropylene housing

product line

Fractogel®

form

prepacked column

parameter

≤8 bar pressure
~240 cm/hr max. flow rate

column I.D. × L

5 mm × 10 mm

volume

0.2 mL

matrix active group

Methacrylate

mean particle size

40-90 μm

capacity

~60 mL, resin binding capacity

application(s)

recombinant protein

separation technique

affinity

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

5 x 10 mm

Features and Benefits

RoboColumn Fractogel® Chelate (M) 0.2 ml enables users to:

  • Efficiently purify histidine-tagged recombinant proteins
  • Coordinate metal ions to non-bonding electron pairs of the amino acid side chains and separate peptides
  • Ensure timely and successful startup and operation
  • Minimize hydrophobic interactions due to the tentacles technology, resulting in superior selectivity and recovery
  • Handle multiple CIP and packing cycles with the chemically and physically robust bead, leading to a longer resin lifetime

Legal Information

FRACTOGEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service