Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12-460

Sigma-Aldrich

Dimethyl-Histone H3 (Lys4) Peptide, biotin conjugate

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes.

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$434.00

$434.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$434.00

About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.41

$434.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

mol wt

Mw 2751 Da

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... H3F3B(3021)

Application

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes.

Quality

Routinely evaluated as a substrate for histone-modifying enzymes.

Physical form

Lyophilized powder

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antimicrobial activity and protease stability of peptides containing fluorinated amino acids.
He Meng,Krishna Kumar
Journal of the American Chemical Society null
Lian-Mei Tan et al.
The Plant cell, 32(7), 2178-2195 (2020-05-03)
Chromatin remodeling and histone modifications are important for development and floral transition in plants. However, it is largely unknown whether and how these two epigenetic regulators coordinately regulate the important biological processes. Here, we identified three types of Imitation Switch

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service