Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.24542

Millipore

Nicotinamide adenine dinucleotide free acid

for biochemistry NAD

Synonym(s):

Nicotinamide adenine dinucleotide free acid, NAD free acid, DPN

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$98.80
5 G
$351.00
25 G
$1,260.00

$98.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$98.80
5 G
$351.00
25 G
$1,260.00

About This Item

Empirical Formula (Hill Notation):
C21H27N7O14P2
CAS Number:
Molecular Weight:
663.43
MDL number:
UNSPSC Code:
12352107
EC Index Number:
200-184-4
NACRES:
NB.21

$98.80


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

solid

pH

2.5-3.0 (20 °C, 10 g/L in H2O)

mp

140-142 °C

solubility

10 g/L

bulk density

200‑300 kg/m3

storage temp.

2-8°C

SMILES string

[P](=O)([O-])(O[P](=O)(OCC4OC(C(C4O)O)[n+]5cc(ccc5)C(=O)N)O)OCC1OC(C(C1O)O)[n]2c3ncnc(c3nc2)N

InChI

1S/C21H27N7O14P2/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33/h1-4,7-8,10-11,13-16,20-21,29-32H,5-6H2,(H5-,22,23,24,25,33,34,35,36,37)

InChI key

BAWFJGJZGIEFAR-UHFFFAOYSA-N

Application


  • Development of a Highly Selective NAD(+) -Dependent Glyceraldehyde Dehydrogenase and its Application in Minimal Cell-Free Enzyme Cascades: This research presents the development of an NAD+-dependent enzyme crucial for bioindustrial applications, demonstrating its utility in enzyme cascades for biosynthesis processes, which could enhance the efficiency of biotechnological production systems (DOI: 10.1002/css).

Analysis Note

Assay (β-NAD, enzymatically with ADH; calc. on anhydrous substance): ≥ 95 %
Assay (spectrophotometrically): ≥ 95.0 %
Identity (IR-spectrum): passes test
Appearance: White to almost white, fine powder eventuel slightly yellowish.
pH (10 g/l, water): 2.5 - 3.0
Absorption maximum λmax. (buffer pH 7.0): 259 - 261 nm
Spec. Absorptivity A1%/1cm (λmax.; 0,01 g/l; buffer pH 7.0; calc. on anhydrous substance): 255 - 270
Water (according to Karl Fischer): ≤ 7.0 %

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service