Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.24518

Supelco

Bathocuproinedisulfonic acid disodium salt

GR for analysis

Synonym(s):

Bathocuproinedisulfonic acid disodium salt, 2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline, 2,9-Dimethyl-4,7-diphenyl-1,10-phenanthrolinedisulfonic acid disodiumsalt, 4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid disodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$1,660.00

$1,660.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$1,660.00

About This Item

Empirical Formula (Hill Notation):
C26H18N2Na2O6S2
CAS Number:
Molecular Weight:
564.54
MDL number:
UNSPSC Code:
12352107
EC Index Number:
258-111-7
NACRES:
NA.21

$1,660.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

solid

pH

10 (20 °C, 10 g/L in H2O)

mp

300 °C

bulk density

190 kg/m3

storage temp.

15-25°C

SMILES string

[Na+].[Na+].[S](=O)(=O)([O-])c1c(nc2c3nc(c(c(c3ccc2c1c5ccccc5)c4ccccc4)[S](=O)(=O)[O-])C)C

InChI

1S/C26H20N2O6S2.2Na/c1-15-25(35(29,30)31)21(17-9-5-3-6-10-17)19-13-14-20-22(18-11-7-4-8-12-18)26(36(32,33)34)16(2)28-24(20)23(19)27-15;;/h3-14H,1-2H3,(H,29,30,31)(H,32,33,34);;/q;2*+1/p-2

InChI key

RNGKZLRAVYPLJC-UHFFFAOYSA-L

Analysis Note

Identity (IR-spectrum): conforms
Identity (UV/VIS-Spectrum): conforms
Absorption maximum λmax. (Buffer pH 5.0; as copper complex): 477 - 485 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.05 g/l; buffer pH 5.0; as copper complex): ≥ 90
Water (according to Karl Fischer): ≤ 10 %
Suitability for copper determination: conforms

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service