Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

123850

Sigma-Aldrich

AKR1C1 Inhibitor, 5-PBSA

The AKR1C1 Inhibitor, 5-PBSA, also referenced under CAS 4906-68-7, controls the biological activity of AKR1C1.

Synonym(s):

AKR1C1 Inhibitor, 5-PBSA, Aldo-Keto Reductase family 1 member C1 Inhibitor, 5-PBSA, 3-Bromo-5-phenylsalicylic acid, 20α-Hydroxysteroid Dehydrogenase Inhibitor, 5-PBSA

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$279.00

$279.00


Please contact Customer Service for Availability


Select a Size

Change View
25 MG
$279.00

About This Item

Empirical Formula (Hill Notation):
C13H9BrO3
CAS Number:
Molecular Weight:
293.11
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$279.00


Please contact Customer Service for Availability

Quality Level

Assay

≥94% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

wet ice

storage temp.

2-8°C

SMILES string

OC1=C(C(O)=O)C=C(C2=CC=CC=C2)C=C1Br

InChI

1S/C13H9BrO3/c14-11-7-9(8-4-2-1-3-5-8)6-10(12(11)15)13(16)17/h1-7,15H,(H,16,17)

InChI key

XVZSXNULHSIRCQ-UHFFFAOYSA-N

General description

A cell-permeable salicylic acid compound that acts as a highly potent, active-site directed and substrate competitive AKR1C1 inhibitor (Ki = 4 nM) with moderate to excellent selectivity over AKR1C2, AKR1C3 and AKR1C4 (Ki = 0.087, 4.2 and 18.2 µM, respectively). Shown to inhibit the metabolism of progesterone by AKR1C1 (IC50 = 460 nM in AKR1C1 overexpressing BAECs).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

El-Kabbani, O., et al. 2011. J. Steroid Biochem. Mol. Biol.125, 105.
El-Kabbani, O., et al. 2009. J. Med. Chem.52, 3259.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service