Skip to Content
MilliporeSigma
All Photos(1)

Documents

12-347

Sigma-Aldrich

Histone H4 Peptide

Histone H4 Peptide primarily used in Histone Acetyltransferase Assays (HAT) and Western Blotting.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.41

Quality Level

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (histone acetyltransferase (HAT))
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Quality

Routinely evaluated by Immunoblot Analysis. This peptide can be used as a standard for antibodies specific for the N-terminus of histone H4. It can also be chemically acetylated with [3H]CH3COONa using the HDAC Assay Kit (17-281). The radiolabeled peptide was used to assay HDAC activity in a HeLa nuclear extract (12-309).

Physical form

Lyophilized powder

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C A Mizzen et al.
Cell, 87(7), 1261-1270 (1996-12-27)
The transcription initiation factor TFIID is a multimeric protein complex composed of TATA box-binding protein (TBP) and many TBP-associated factors (TAF(II)s). TAF(II)s are important cofactors that mediate activated transcription by providing interaction sites for distinct activators. Here, we present evidence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service