Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

120497

Supelco

AUS Adapter for Power Supply NOVA

for use with SPECTROQUANT®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116

compatibility

for use with SPECTROQUANT®

shipped in

ambient

storage temp.

15-25°C

General description

Spectroquant®

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service