119114
Luciferase Inhibitor II
The Luciferase Inhibitor II, also referenced under CAS 10205-56-8, controls the biological activity of Luciferase.
Synonym(s):
Luciferase Inhibitor II, 4-(1,3-Benzothiazol-2-yl)-N,N-dimethylaniline
Select a Size
$181.00
Select a Size
About This Item
$181.00
Recommended Products
Quality Level
Assay
≥95% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
pale green
solubility
DMSO: 0.5 mg/mL
ethanol: 3 mg/mL
shipped in
ambient
storage temp.
2-8°C
SMILES string
[s]1c2c(nc1c3ccc(cc3)N(C)C)cccc2
InChI
1S/C15H14N2S/c1-17(2)12-9-7-11(8-10-12)15-16-13-5-3-4-6-14(13)18-15/h3-10H,1-2H3
InChI key
HYKGLCSXVAAXNC-UHFFFAOYSA-N
General description
Packaging
Warning
Reconstitution
Other Notes
Auld, D.S., et al. 2008. J. Med. Chem.51, 2372.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service