Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.18386

Supelco

Aluminum Test Kit, colorimetric

0.10-6 mg/L (Al), MQuant®

Synonym(s):

Aluminium Visual Test Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Aluminium Test, colorimetric, 0.10-6 mg/L (Al), for use with MCOLORTEST®

specific analyte(s)

aluminium

Quality Level

measuring range

0.10-6 mg/L (Al)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color-disk comparator 0.10 - 0.20 - 0.35 - 0.50 - 0.75 - 1 - 2 - 3 - 6 mg/l Al MQuant®
The MQuant® Aluminium test (formally MColortest®) measures aluminium in e.g. groundwater and surface water, seawater, drinking water, industrial water, waste water and percolating water.

Method
In weakly acidic, acetate-buffered solution aluminium ions react with chromazurol S to form a blue compound. Due to the intrinsic orange coloration of the reagent blank, the measurement solution is orange-red to red in color. The aluminium concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color disk. The test evaluates the color reaction based on the transmitted light method. So even turbid and colored water samples can be analyzed without further preparation.

The color disk is made of very durable, lightfast plastic, thus suitable for industrial areas and wet environments.
Using the refill pack 1.18452 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system.

The test can be used for the determination of aluminium in
  • Groundwater and surface water, seawater
  • Drinking water and mineral water
  • Waters from aquaculture
  • Boiler and boiler feed water, cooling water
  • Process water
  • Industrial water
  • Wastewater and percolating water
  • Electroplating wastewater
  • Swimming-pool water

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service