Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.18323

Supelco

Silicate (Silicic Acid) Test Refill pack

for use with MQUANT®

Synonym(s):

Silicate Visual Test Kit Refills

Sign Into View Organizational & Contract Pricing

Select a Size

150 TESTS
$208.00

$208.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
150 TESTS
$208.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$208.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

feature

refill

Quality Level

specific analyte(s)

silicate

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

This refill pack contains all the reagents for the MQuant® test kits 1.14792 and 1.14410 for the colorimetric evaluation of silicate. When the reagents of the MQuant® test are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of MQuant® test kits, making it a particularly economical and ecological system.

Method
In sulfuric solution silicate ions react with molybdate ions to form a yellow heteropoly acid. This is reduced to silicomolybdenum blue. The silicate concentration is measured semiquantitatively by visual colorimetric evaluation.

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - Skin Sens. 1

Supplementary Hazards

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service