Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.17104

Millipore

MAS-100 VF® Power supply

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$256.00

$256.00


In StockDetails


Request more information

Select a Size

Change View
1 PKG
$256.00

About This Item

UNSPSC Code:
41122409
NACRES:
NA.74

$256.00


In StockDetails


Request more information

material

aluminum hardware

Agency

non-compendial

feature

electronic

packaging

pkg of 1 kit

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with MAS-100 VF®

shipped in

ambient

storage temp.

no temp limit

General description

Power supply is an additional accessory for MAS-100 VF® air sampler. The portable MAS-100 VF® is designed to efficiently monitor air microbiology in cleanrooms and other controlled environments in which microbiological contamination has a direct impact on product quality, shelf life and deterioration. Like all other MAS air samplers, the MAS-100 VF® is based on the Andersen impaction principle. The power supply has an input voltage of 100 to 240 V AC (47 to 63 Hz, 1.5 to 0.7 A) and output voltage of 24 V DC ± 5% (2.7 A). The cable length varies depending upon the country.

Application

MAS-100 VF® Power supply is an additional accessory. Microbial air sampling devices are extensively used in Pharmaceutical and Personal care industries as well as Food, Beverage and Environmental applications.

Legal Information

MAS-100 VF is a registered trademark of MBV AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service