Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.16740

Supelco

Petroleum benzine boiling range 40−60 GRAD C hypergrade for organic trace analysis

SupraSolv®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12191502
NACRES:
NA.03
technique(s):
GC/MS: suitable
bp:
40-60 °C (≥ 90 (% v/v))
Pricing and availability is not currently available.

description

for trace analysis

Quality Level

product line

SupraSolv®

technique(s)

GC/MS: suitable

impurities

≤0.005% water

( Evapor)

≤3.0 mg/L

color

≤10, Hazen

bp

40-60 °C (≥ 90 (% v/v))

General description

SupraSolv® hypergrade for organic trace analysis is a unique and universal solvent quality which can be employed for all three GC detection methods (ECD, FID and MS). Thanks to the large retention time range, even low-boiling substances can be reliably analyzed.

Analysis Note

GC/ECD (retention range 1,2,4-trichlorobenzene to decachlorobiphenyle, individual signals (lindane standard)) = 2 pg/ml
GC/FID (retention range n-undecane to n-tetracontane, individual signals (n-tetradecane standard)) = 2 ng/ml
GC/MSD (retention range n-undecane to n-tetracontane, scaning area 30 - 600 amu, individual signals (n-tetradecane standard)) = 2 ng/ml

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F

Flash Point(C)

-22 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service