Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.15948

Sigma-Aldrich

Safranine O (C.I. 50240)

for microscopy Certistain®

Synonym(s):

Safranine O (C.I. 50240)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19ClN4
CAS Number:
Molecular Weight:
350.84
UNSPSC Code:
41116130
EC Index Number:
207-518-8

Quality Level

form

solid

composition

Dye, ≥80% spectrophotometric assay

pH

10 (20 °C, 10 g/L in H2O)

solubility

50 g/L

bulk density

400 kg/m3

εmax

1260-1570 at 530-534 nm in 50% ethanol at 0.003 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C20H18N4.ClH/c1-12-10-17-19(11-16(12)22)24(14-6-4-3-5-7-14)18-9-8-15(21)13(2)20(18)23-17;/h3-11,22H,21H2,1-2H3;1H

InChI key

QRYAEWIQIBAZOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Safranin O (C.I. 50240) for microscopy Certistain®, is a dry dye that is used for the preparation of a staining solution for e.g. Gram stain procedures in smears and sections of human origin in bacteriology. The Gram staining allows a fast differentiation of bacteria in Gram-positive and Gram-negative. The mureine structure of the bacteria walls is the basis of the color affinity. Gram-negative bacteria will be counterstained by Gram′s safranin O solution and appear orange. As a Certistain® dye it is chemically quality-certified according to highest standards and strict specifications. As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Dye content (spectrophotometrically): ≥ 80 %
Identity (UV/VIS-Spectrum): passes test
Absorption maximum λ max (ethanol 50 %): 530 - 534 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.003 g/l; ethanol 50 %): 1260 - 1570
TLC-Test: passes test
Loss on drying (110 °C): ≤ 15 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service