Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.15943

Sigma-Aldrich

Methylene blue (C.I.52015)

for microscopy Certistain®

Synonym(s):

Methylene Blue trihydrate, Methylthioninium chloride, 3,7-Bis(dimethylamino)phenothiazinium chloride, Tetramethylthionine chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18ClN3S · 3H2O
CAS Number:
Molecular Weight:
373.90
Beilstein:
3599847
MDL number:
UNSPSC Code:
41116130
EC Index Number:
200-515-2
Pricing and availability is not currently available.

Quality Level

form

solid

potency

1180 mg/kg LD50, oral (Rat)

composition

Dye, ≥82% spectrophotometric assay

pH

3 (20 °C, 10 g/L in H2O)

mp

190 °C (dec.) (lit.)

solubility

50 g/L

density

1.757 g/cm3 at 22 °C

bulk density

400‑600 kg/m3

εmax

2250-2750 at 660-665 nm in 50% ethanol at 0.003 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

SMILES string

[Cl-].CN(C)c1ccc2N=C3C=C\C(C=C3Sc2c1)=[N+](/C)C

InChI

1S/C16H18N3S.ClH/c1-18(2)11-5-7-13-15(9-11)20-16-10-12(19(3)4)6-8-14(16)17-13;/h5-10H,1-4H3;1H/q+1;/p-1

InChI key

CXKWCBBOMKCUKX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Methylene blue (C.I. 52015) for microscopy Certistain® is a dry dye that is used in bacteriology for an overview stain and as counterstaining for Ziehl-Neelsen method in sample material of human origin. Bacteria appear blue. The dye has a further important application in hematology: Methylene blue and eosin are the main components of many staining solutions and dye mixtures. Certistain® is the product group for seleceted high quality dyes that fulfill all quality requirements of IVD and CE registration. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Dye content (spectrophotometrically): ≥ 82 %
Identity
- Chloride:
passes test
- UV/VIS-Spectrum: passes test
Absorption maximum λ max (ethanol 50 %): 660 - 665 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.003 g/l; ethanol 50 %): 2250 - 2750
TLC-Test: passes test
Loss on drying (110 °C): 10 - 15 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service