Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.15931

Sigma-Aldrich

Acridine orange zinc chloride double salt (C.I.46005)

for microscopy Certistain®

Synonym(s):

Acridine orange zinc chloride double salt (C.I.46005)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20Cl3N3Zn
CAS Number:
Molecular Weight:
438.11
MDL number:
UNSPSC Code:
41116130
EC Index Number:
233-353-6

Quality Level

form

solid

composition

Dye, ≥50% spectrophotometric assay

pH

3.9 (20 °C, 10 g/L in H2O)

mp

165 °C

solubility

28 g/L

bulk density

200 kg/m3

εmax

≥1120 at 491-495 nm in 50% ethanol at 0.005 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C17H19N3.3ClH.Zn/c1-19(2)14-7-5-12-9-13-6-8-15(20(3)4)11-17(13)18-16(12)10-14;;;;/h5-11H,1-4H3;3*1H;/q;;;;+2/p-2

InChI key

VADJQOXWNSPOQA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Acridine orange zinc chloride double salt (C.I. 46005) for microscopy Certistain®, is a< dry dye that is used for the preparation of a staining solution for fluorescence staining for differential staining of DNA and RNA in sample material of human origin. As a Certistain® dye it is chemically quality-certified according to highest standards and strict specifications. As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Dye content (spectrophotometrically): ≥ 50% C₁₇H₂₀ClN₃
Identity
- chloride:
passes test
- UV/VIS-Spectrum: passes test
- Zinc: passes test
Absorption maximum λ max (ethanol 50 %): 491 - 495 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l; ethanol 50 %): ≥ 1120
TLC-Test: passes test
Loss on drying (110 °C): ≤ 8 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service