Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.15927

Sigma-Aldrich

Ponceau S (C.I. 27195)

for microscopy Certistain®

Synonym(s):

Ponceau S (C.I. 27195)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H12N4Na4O13S4
CAS Number:
Molecular Weight:
760.57
MDL number:
UNSPSC Code:
41116130
EC Index Number:
228-319-2

Quality Level

form

solid

composition

Dye, ≥80% spectrophotometric assay

pH

9.0 (20 °C, 10 g/L in H2O)

solubility

10 g/L

bulk density

660 kg/m3

εmax

460-600 at 517-523 nm in water at 0.01 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C22H16N4O13S4.4Na/c27-22-20(43(37,38)39)10-12-9-16(41(31,32)33)6-7-17(12)21(22)26-25-18-11-14(3-8-19(18)42(34,35)36)24-23-13-1-4-15(5-2-13)40(28,29)30;;;;/h1-11,25H,(H,28,29,30)(H,31,32,33)(H,34,35,36)(H,37,38,39);;;;/q;4*+1/p-4/b24-23?,26-21-;;;;

InChI key

LZYGEJPERQQCIY-STNZDNLRSA-J

Looking for similar products? Visit Product Comparison Guide

General description

The dry dye Ponceau S (C.I. 27195) for microscopy Certistain®, is used for the preparation of a staining solution for staining connective tissue in histological sections of human origin.
As Certistain® dye it is chemically quality-certified according to highest standards and strict specifications. As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Dye content (spectrophotometrically): ≥ 80 %
Identity
- sodium:
passes test
- UV/VIS-Spectrum: passes test
Absorption maximum λ max (water): 517 - 523 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.01 g/l; water): 460 - 600
TLC-Test: passes test
Loss on drying (110 °C): ≤ 10 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service