Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.15533

Millipore

CULTURA mini incubator

115 V, Bench-top incubator, suitable for incubation of contact slides, dipslides, and Petri dishes

Synonym(s):

Incubator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
42281526
NACRES:
NB.14

description

For use with MULTIRACK (1.13312)

packaging

pkg of (1 unit)

application(s)

food and beverages

storage temp.

room temp

General description

CULTURA mini incubator allows easy incubation of microbiological samples like Envirocheck Contact slides, dipslides, and Petridishes. It brings a flexible working environment by reducing the necessary workspace. It provides a tray with holes ready for the Envirocheck Contact slide tubes. It is robust, easy-to-clean, and allows various incubation temperatures.

Application

The CULTURA Mini Incubator is used for the incubation of contact slides and dipslides, as well as common Petri dishes for detecting bacteria, fungi and yeast. The incubator can be used to monitor drinking water, process water, and swimming pool water, cooling, lubricant and industrial liquid, in food bacteriology and hygiene control, as well as in medical and dental diagnostics.

Features and Benefits

  • Bench-top system
  • Light-weighted

Legal Information

ENVIROCHECK is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service