Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1150-OP

Millipore

Acrylamide

OmniPur® Grade, >=99.9%, Suitable for polyacrylamide gel electrophoresis

Synonym(s):

OmniPur® Acrylamide, Propenamide, Acrylic Acid Amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCONH2
CAS Number:
MDL number:
UNSPSC Code:
41105319

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear colorless

foreign activity

Protease, none detected
RNAse, none detected

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)

InChI key

HRPVXLWXLXDGHG-UHFFFAOYSA-N

General description

Suitable for polyacrylamide gel electrophoresis.

Specifications:
Assay: ≥99.9%

Conductivity (40%, water): ≤5 µmho
Free Acrylic Acid: ≤0.001%
Iron (Fe): ≤0.0001%
Lead (Pb): ≤0.0001%
Melting point: 84-86°C
pH (10% in 100 mM NaCl, , 25°C): 5.5-6.5
Protease: None detected
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service