Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.14901

Supelco

Flat-bottomed tubes with screw caps

long tubes, MQuant®

Sign Into View Organizational & Contract Pricing

Select a Size

12 UNITS
$148.00

$148.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
12 UNITS
$148.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$148.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Product Name

Flat-bottomed long tubes with screw caps, for use with MQUANT®

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

We offers you useful accessories. These long flat-bottomed tubes can be used for our MQuant® test kits with color card comparator (formally MColortest®). The screw caps are included.
for MQuant® tests with color card comparator MQuant®

Analysis Note

Application test: conforms

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Photometric assessment of bromate in drinking water samples using Spectroquant® photometer and test kits, along with 3,3'-dimethylnaphthidine and potassium iodide.

Protocols

Photometric determination of phosphate ions in water with ammonium molybdate using Spectroquant® test kits and a UV/VIS spectrophotometer.

Questions

  1. Posso sapere il diametro dei tubi e se posso utilizzarli per la determinazione visiva del colore e dell'opalescenza?

    1 answer
    1. The outer diameter is 20 mm. This clear tube is used for MQuant colorimetric tests and should work for visual determinations of color and opalescence.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service