Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.14839

Supelco

Boron Test, photometric

photometric, 0.050-0.800 mg/L (B), Spectroquant®

Sign Into View Organizational & Contract Pricing

Select a Size

60 TESTS
$500.00

$500.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
60 TESTS
$500.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$500.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Product Name

Boron Test, photometric, 0.050-0.800 mg/L (B), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 60 tests

specific analyte(s)

boron

measuring range

0.050-0.800 mg/L (B)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Rosocyanine)

storage temp.

15-25°C

General description

Method: photometric 0.050 - 0.800 mg/l B Spectroquant®

The method is analogous to EPA 212.3, APHA 4500-B B, and withdrawn ASTM D3082-15.

This Spectroquant® Boron Reagent Test allows the accurate quantification of the boron content in aqueous samples.

Method applied: In acidic solution borate ions react with a diol to form chelates that are selectively extracted. In the extract the chelates react with curcumin to form a red complex which is determined photometrically.

The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.

All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1A - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service