Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.14800

Supelco

Cyanide Test

photometric, 0.002-0.500 mg/L (CN-), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116

product line

Spectroquant®

Quality Level

specific analyte(s)

cyanide

reaction suitability

reaction type: Koenigs-Knorr Reaction

measuring range

0.002-0.500 mg/L (CN-)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric

storage temp.

15-25°C

General description

(free cyanide) Method: Photometric, 0.002 - 0.500 mg/L (CN-), Spectroquant®

The method is analogous to EPA 335.2, APHA 4500-CN- E, ASTM D2036-09 D, ISO 6703, and DIN 38405-13.

This Spectroquant® Cyanide Reagent Test allows the accurate quantification of the free cyanide content in aqueous samples.

Method applied: Cyanide ions react with a chlorinating agent to form cyanogen chloride. In a follow-up reaction with 1,3-dimethylbarbituric acid in the presence of pyridine a violet dye (König reaction) is formed which is determined photometrically.

The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.

All our Cell and Reagent Test Kits are equipped with the unique Live ID which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service