Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.14652

Supelco

Total Hardness Test Kit, titrimetric

titrimetric, for use with MCOLORTEST®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Total Hardness Test in freshwater, titrimetric, for use with MCOLORTEST®

specific analyte(s)

calcium, magnesium

Quality Level

compatibility

for use with MCOLORTEST®

detection method

titrimetric

storage temp.

15-25°C

General description

Method: titrimetric with dropping bottle MColortest®
The test measures the content of salts of the alkaline earth metals calcium, magnesium, strontium and barium in freshwater. For the determination the titrant is added dropwise to the sample until the added indicator changes its color.
This MQuant® Total Hardness test (formally MColortest®) is suitable for the titrimetric determination using a convenient dropping bottle.

Method
Calcium and magnesium ions react with an indicator to form a red complex compound. The indicator is released from this compound by titration with a solution of ethylenedinitrilotetraacetic acid disodium salt dihydrate (Titriplex® III). At the titration end-point the color changes to green. The total hardness is determined from the consumption of titration solution.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Titriplex is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service