Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.14567

Supelco

Replacement filter

for use with Extrelut® NT 20 HDPE columns

Sign Into View Organizational & Contract Pricing

Select a Size

50 UNITS
$347.00

$347.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 UNITS
$347.00

About This Item

UNSPSC Code:
41115711
NACRES:
NB.21

$347.00


Please contact Customer Service for Availability

Request a Bulk Order

technique(s)

supported liquid extraction (SLE): suitable

Quality Level

compatibility

for use with Extrelut® NT 20 HDPE columns

storage temp.

no temp limit

General description

for EXtrelut® NT 20 HDPE columns

Analysis Note

purity test (TLC): conforms

Legal Information

Extrelut is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Simplify your liquid-liquid extractions with our emulsion-free supported-liquid extraction (SLE) single step solution.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service