Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.14500

Supelco

Formaldehyde Test, photometric

photometric, 0.10-8.00 mg/L (HCHO), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Formaldehyde Cell Test, photometric, 0.10-8.00 mg/L (HCHO), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

specific analyte(s)

formaldehyde

measuring range

0.10-8.00 mg/L (HCHO)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric

storage temp.

15-25°C

General description

Method: photometric 0.10 - 8.00 mg/l HCHO Spectroquant®


This Spectroquant® Formaldehyde Cell Test allows the accurate quantification of the formaldehyde content in aqueous samples such as disinfectants, preservatives, or process wastewater.


Method applied: In sulfuric solution formaldehyde reacts with chromotropic acid to form a violet dye which is determined photometrically.


The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.


All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Application


  • Effect of formaldehyde exposure on phytochemical content and functional activity of Agaricus bisporus (Lge.) Sing: This research highlights the application of formaldehyde detection assays in studying the impact of environmental toxins on biological materials, which is critical for assessing chemical exposure and material safety in both laboratory and industrial settings (Kutluer, 2024).

  • VOC data-driven evaluation of vehicle cabin odor: from ANN to CNN-BiLSTM: This study employs advanced formaldehyde detection techniques, integral for environmental toxin testing in automotive manufacturing, ensuring cabin air quality and safety standards are met (Tian et al., 2024).

  • Graphene and metal-organic framework hybrids for high-performance sensors for lung cancer biomarker detection supported by machine learning augmentation: While primarily for biomarker detection, the sensitivity of these sensors could be adapted for high-sensitivity formaldehyde assays in biotech and pharma research, showcasing the inter-disciplinary application of formaldehyde assays (Tran et al., 2024).

  • A metal-organic framework and quantum dot-based ratiometric fluorescent probe for the detection of formaldehyde in food: This study represents a direct application of formaldehyde cell tests in food safety, illustrating the role of formaldehyde detection assays in ensuring public health and compliance with safety regulations (Chen et al., 2024).

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Preparation of a standard solution for Formaldehyde

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service