Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.14411

Supelco

Sulfate Test Kit, colorimetric

25-300 mg/L (SO₄²⁻), MQuant®

Synonym(s):

Sulfate Visual Test Kit

Sign Into View Organizational & Contract Pricing

Select a Size

90 TESTS
$305.00

$305.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
90 TESTS
$305.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$305.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Product Name

Sulfate Test, colorimetric, 25-300 mg/L (SO42-), for use with MCOLORTEST®

specific analyte(s)

sulfate

Quality Level

measuring range

25-300 mg/L (SO42-)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color card and comparator block with short test tubes 25 - 50 - 80 - 110 - 140 - 200 - 300 mg/l SO MQuant®
The MQuant® Sulfate test (formally MColortest®) is used for the determination of sulfate in:
  • Groundwater and surface water
  • Drinking water and mineral water
  • Wastewater
This test is not suited for seawater.

Method
Sulfate ions react with barium iodate, liberating iodate ions in the process. These oxidize tannin to a brown-red compound. The sulfate concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color card. The unique brilliance of the print and their fine color graduation allow precise analyses, even at the lower end of the concentration range.
Using the refill pack 1.18467 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system. 

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Ox. Liq. 2 - Repr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service