Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.13713

Supelco

1,2-Dichloroethane

for liquid chromatography LiChrosolv®

Synonym(s):

Ethylene chloride, Ethylene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2Cl
CAS Number:
Molecular Weight:
98.96
Beilstein:
605264
MDL number:
UNSPSC Code:
12191502
EC Index Number:
203-458-1
Assay:
≥99.8% (GC)
technique(s):
HPLC: suitable
bp:
83 °C (lit.)
vapor pressure:
87 mmHg ( 25 °C)

vapor density

3.4 (20 °C, vs air)

Quality Level

vapor pressure

87 mmHg ( 25 °C)

product line

LiChrosolv®

grade

isocratic

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

775 °F

potency

670 mg/kg LD50, oral (Rat)
2800 mg/kg LD50, skin (Rabbit)

expl. lim.

15.6 %

technique(s)

HPLC: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.02% Water

evapn. residue

≤2.0 mg/L

transmittance

240 nm, ≥85%
245 nm, ≥90%
270 nm, ≥99%

refractive index

n20/D 1.444 (lit.)

bp

83 °C (lit.)

mp

−35 °C (lit.)

transition temp

flash point 13 °C

density

1.256 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

ClCCCl

InChI

1S/C2H4Cl2/c3-1-2-4/h1-2H2

InChI key

WSLDOOZREJYCGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.

Specifications

Saturation Concentration in Air: 350 g/m3 (20 °C) Air

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Evaporation Number: 4.1
Purity (GC): ≥ 99.8 %
Identity (IR): conforms
Evaporation residue: ≤ 2.0 mg/l
Water: ≤ 0.02 %
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Transmission (at 240 nm): ≥ 85 %
Transmission (at 245 nm): ≥ 90 %
Transmission (from 270 nm): ≥ 99 %
Filtered by 0.2 µm filter

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service