Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.12936

Supelco

Oxidizing decomposition reagent

for compounds containing P and N

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352402
NACRES:
NA.21

Quality Level

form

solid

pH

11 (20 °C, 10 g/L in H2O)

bulk density

800‑900 kg/m3

storage temp.

15-25°C

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service