Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.12298

Supelco

Triton X-100

for gas chromatography

Synonym(s):

Octoxynol, (p-tert-Octylphenoxy)polyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol, Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, t-Octylphenoxypolyethoxyethanol, Polyethylene glycol tert-octylphenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
t-Oct-C6H4-(OCH2CH2)xOH, x= 9-10
CAS Number:
Beilstein:
2315025
MDL number:
UNSPSC Code:
12352104

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

form

liquid

potency

1900-5000 mg/kg LD50, oral (Rat)
>3000 mg/kg LD50, skin (Rabbit)

mol wt

micellar avg mol wt 80,000
average mol wt 625

aggregation number

100-155

technique(s)

gas chromatography (GC): suitable

pH

5.0-8.0 (20 °C, 10 g/L in H2O)

CMC

0.2-0.9 mM (20-25°C)

bp

>200 °C/1013 hPa

transition temp

flash point 251 °C
cloud point 65 °C
pour point ~7 °C

density

1.07 g/cm3 at 20 °C

HLB

13.5

storage temp.

2-30°C

SMILES string

CC(C)(C)CC(C)(C)c1ccc(OCCOCCOCCOCCOCCOCCOCCO)cc1

InChI

1S/C28H50O8/c1-27(2,3)24-28(4,5)25-6-8-26(9-7-25)36-23-22-35-21-20-34-19-18-33-17-16-32-15-14-31-13-12-30-11-10-29/h6-9,29H,10-24H2,1-5H3

InChI key

HNLXNOZHXNSSPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application



  • Regression analysis for the determination of microplastics in sediments using differential scanning calorimetry.: This study utilized sea sand as a matrix to investigate the presence of microplastics in sediment samples through differential scanning calorimetry (Schirrmeister et al., 2024) (Schirrmeister et al., 2024).


  • Combination of Sea Sand Disruption Method and Ion-Pair Solid-Phase Extraction for Effective Isolation and Purification of Chlorogenic Acid from Plants Prior to the HPLC Determination.: The research developed a novel method combining sea sand disruption and ion-pair solid-phase extraction to enhance the isolation and purification of chlorogenic acid from plant matrices (Wianowska, 2022) (Wianowska, 2022).


  • Vortex-assisted matrix solid-phase dispersion: An eco-friendly alternative for the determination of halogens in edible seaweed.: Sea sand was employed in vortex-assisted matrix solid-phase dispersion, presenting an eco-friendly technique for analyzing halogens in seaweed (de Melo Malinowski et al., 2022) (de Melo Malinowski et al., 2022).


  • Matrix solid phase dispersion-assisted BCR sequential extraction method for metal partitioning in surface estuarine sediments.: This study used sea sand in matrix solid-phase dispersion to improve the extraction and partitioning analysis of metals in estuarine sediments (Martínez-Fernández et al., 2011) (Martínez-Fernández et al., 2011).




Biochem/physiol Actions

Widely used non-ionic surfactant for recovery of membrane components under mild non-denaturing conditions.

Analysis Note

Identity (IR-spectrum): passes test
McReynolds constant (Benzene): 190 - 250
McReynolds constant (Butanol): 370 - 420
McReynolds constant (Nitropropane): 365 - 405
Temperaturestability: loss in weight between 270 and 290 °C (thermogravimetry): ≤ 2 %
Water (according to Karl Fischer): ≤ 0.2

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

483.8 °F - closed cup

Flash Point(C)

251 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Learn about the sample preparation and photometric determination of non-ionic surfactants in Extran® rinse solutions (MA 01, MA 02, MA 05, AP 22) using Spectroquant® test kits and spectrophotometers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service