Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.11805

Supelco

2,2,2-Trifluoro-N-methyl-N-(trimethylsilyl)acetamide

for gas chromatography

Synonym(s):

2,2,2-Trifluoro-N-methyl-N-(trimethylsilyl)acetamide, MSTFA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12F3NOSi
CAS Number:
Molecular Weight:
199.25
MDL number:
UNSPSC Code:
12352103
EC Index Number:
246-331-6

Assay

≥95.0% (GC)

form

liquid

bp

130-132 °C/1013 hPa

transition temp

flash point 25 °C

density

1.085 g/cm3

storage temp.

2-30°C

InChI

1S/C6H12F3NOSi/c1-10(12(2,3)4)5(11)6(7,8)9/h1-4H3

InChI key

MSPCIZMDDUQPGJ-UHFFFAOYSA-N

Application


  • CO2 Capture: 1-Methyl-2-pyrrolidinone is used in bench-scale CO2 capture processes due to its excellent solvent properties. Recent research has demonstrated the effectiveness of NMP in combination with 2-(ethylamino) ethanol and water in capturing CO2 efficiently. This application is crucial for developing sustainable methods to reduce carbon emissions from industrial sources (Qi et al., Fuel, 2023).


Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service