Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.11674

Sigma-Aldrich

Hemacolor® Rapid staining of blood smear

suitable for hematology, staining set for microscopy

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
$178.00

$178.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 SET
$178.00

About This Item

UNSPSC Code:
41116121
NACRES:
NA.21

$178.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

liquid

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

application(s)

clinical testing
diagnostic assay manufacturing
histology

hematology

storage temp.

15-25°C

General description

The Hemacolor® Rapid staining of blood smear - staining set for microscopy, is used for human-medical cell diagnosis and serves the purpose of the hematological and clinico-cytological investigation of sample material of human origin, for example smears of whole blood and bone marrow.
The Hemacolor® Rapid staining of blood smear staining set combines the brilliance and reproducibility of the Pappenheim staining with the time-advantage of a rapid stain. The Hemacolor® Rapid staining of blood smear staining set contains a fixing solution, one red (eosin) and one blue (azur) staining solution and phosphate buffer tablets pH 7.2 according to Weise (Product number 1.09468). The staining solutions are separately applied, which results in a high degree of stability. The conventionally appearing precipitates in the hematological staining solutions, could be avoided in this kit. In addition to blood and bone marrow smears, Hemacolor® Rapid staining of blood smear can be used in clinical cytology, e.g. for staining urine, sputum, FNAB, effusions and lavages.
A kit contains safe 100-ml PE-bottles and three buffer tablets. The package is sufficient for 50 - 200 applications. The buffer tablets acc. to Weise pH 7.2 (Product number 1.09468) are also available in larger pack sizes, thus, they can be useful for higher demands. The three solutions needed for staining are all available individually as:

Hemacolor® Rapid staining of blood smear solution 1, fixing solution (Product number 1.11955)
Hemacolor® Rapid staining of blood smear solution 2, colour reagent red (Product number 1.11956)
Hemacolor® Rapid staining of blood smear solution 3, colour reagent blue(Cat. no. 111957)

For more details, please see instructions for use (IFU). This product is registered as IVD and CE marked.

Analysis Note

Suitability for microscopy (Blood smear): passes test
Erythrocytes: pink to brownish
Nuclei: red-violet
Eosinophilic granules: red to red-brown
Neutrophilic granules: light violet
Lymphocyte cytoplasm: blue

Legal Information

HEMACOLOR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Kann man mit diesem Kit Auch Blutausstriche herstellen, wo das Blut vorher in EDTA Röhrchen gelagert wurde?

    1 answer
    1. Yes, the Hemacolor reagents are suitable for use with samples anticoagulated with EDTA. If the sample is not anticoagulated, the smears must be made immediately upon the blood being drawn. These reagents are not suitable for use with samples anticoagulated with heparin or sodium citrate. Smears should be prepared from fresh samples when prepared from anticoagulated blood with EDTA.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service