Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.11474

Millipore

1,4-Dithiothreitol

for biochemistry

Synonym(s):

1,4-Dithiothreitol, DTT, Cleland’s reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H3OHSH)2
CAS Number:
Molecular Weight:
154.25
MDL number:
UNSPSC Code:
12352104
EC Index Number:
222-468-7
NACRES:
NB.21

grade

for biochemistry

Quality Level

Assay

≥99% (iodometric)

form

solid

potency

400 mg/kg LD50, oral (Rat)

reaction suitability

reagent type: reductant

pH

5.1 (20 °C, 10 g/L in H2O)

mp

40-43 °C

transition temp

flash point >110 °C

bulk density

300 kg/m3

storage temp.

2-8°C

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4-/m0/s1

InChI key

VHJLVAABSRFDPM-IMJSIDKUSA-N

Application


  • Drug delivery using reduction-responsive hydrogel based on carboxyethyl-succinoglycan with highly improved rheological, antibacterial, and antioxidant properties.: This study explores the application of 1,4-Dithiothreitol in developing reduction-responsive hydrogels, highlighting significant enhancements in rheological, antibacterial, and antioxidant characteristics, suitable for drug delivery systems (Kim et al., 2024).

  • Preparation and properties of hydrogel photonic crystals assembled by biodegradable nanogels.: Research discusses the integration of 1,4-Dithiothreitol in the assembly of hydrogel photonic crystals using biodegradable nanogels, emphasizing their potential in creating novel photonic materials (Zhang et al., 2024).

  • Optimized total RNA isolation from bovine sperm with enhanced sperm head lysis.: The paper highlights the use of 1,4-Dithiothreitol in optimizing RNA isolation techniques from bovine sperm, which enhances sperm head lysis, crucial for reproductive biology studies (Tiwari et al., 2024).

  • Preparation of a Convertible Spacer Containing a Disulfide Group for Versatile Functionalization of Oligonucleotides.: Discusses the synthesis of a convertible spacer containing a disulfide group using 1,4-Dithiothreitol, enabling versatile functionalization of oligonucleotides for therapeutic and diagnostic applications (Pontarelli et al., 2023).

Analysis Note

Assay (iodometric): ≥ 99 %
Identity (IR-spectrum): passes test
Melting point: 40 - 43 °C
Heavy metals (as Pb): ≤ 10 ppm
Fe (Iron): ≤ 5 ppm

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service