Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.10690

Millipore

GranuCult® plus DEV Lactose peptone broth

for coliforms, suitable for microbiology

Synonym(s):

Coliform enrichment broth, Lactose Peptone Broth DEV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74
Pricing and availability is not currently available.

form

medium granules (dehydrated (DCM))

Quality Level

packaging

pkg of 500 g

manufacturer/tradename

GranuCult® plus

storage condition

protect from light (Keep dry and tightly closed)

technique(s)

microbiological culture: suitable

color

violet

pH

7.0 (37 °C, 25 g/L in H2O, after autoclaving)

solubility

25 g/L at 100 °C, clear

bulk density

700 kg/m3

application(s)

food and beverages
water monitoring

microbiology

storage temp.

15-25°C

suitability

coliforms

General description

GranuCult® plus DEV Lactose peptone broth is suited for the enrichment and determination of the titer of coliform bacteria in the bacteriological analysis of water according to the German standard methods for the examination of water and the previous German regulations from 1990.Casein peptone and soya peptone provide nitrogen, vitamins, minerals, and amino acids for growth, while sodium chloride ensures osmotic balance. Lactose is a carbohydrate source. The pH indicator bromocresol purple is yellow below pH 5.3 (acidic) and purple above pH 6.7 (alkaline). The fermentation of lactose and the consequent acid production changes the color of the medium to yellow.

Application

GranuCult® plus DEV Lactose peptone broth is suitable for the enrichment of coliforms in the bacteriological analysis of water.

Features and Benefits

  • GranuCult® offers superior granulated culture media.
  • Safe and sustainable due to reduced risks associated with fine dust and toxic substance inhalation, resulting in a safer work environment.
  • Excellent wettability, solubility, and free-flowing properties
  • Convenient, with minimal component separation and clumping, even under warm or humid conditions.
  • High batch-to-batch reproducibility
  • Prolonged shelf life of up to five years
  • High number of test strains exceeding all regulatory demands
  • Granulation technology allows many supplements to be included, with no need to add these separately

Analysis Note

Appearance (clearness): clear
Appearance (colour): violet
pH-value (25 °C): 7.0 - 7.4
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): good to very good
Growth (Klebsiella pneumoniae ATCC 13883 (WDCM 00097)): good to very good
Growth (Salmonella typhimurium ATCC 14028 (WDCM 00031)): good to very good
Growth (Aeromonas hydrophila ATCC 7966 (WDCM 00063)): medium to very good
Growth (Enterococcus faecalis ATCC 11700): medium to very good
Colour change to yellow (Escherichia coli ATCC 25922 (WDCM 00013)): +
Colour change to yellow (Klebsiella pneumoniae ATCC 13883 (WDCM 00097)): +
Colour change to yellow (Salmonella typhimurium ATCC 14028 (WDCM 00031)): -
Colour change to yellow (Aeromonas hydrophila ATCC 7966 (WDCM 00063)): -
Colour change to yellow (Enterococcus faecalis ATCC 11700): +/-
Gas (Escherichia coli ATCC 25922 (WDCM 00013)): +
Gas (Klebsiella pneumoniae ATCC 13883 (WDCM 00097)): +
Gas (Salmonella typhimurium ATCC 14028 (WDCM 00031)): -
Gas (Aeromonas hydrophila ATCC 7966 (WDCM 00063)): -
Gas (Enterococcus faecalis ATCC 11700): -
Incubation: 24 hrs.; 35 °C; aerobic.

Other Notes

  • Do not use clumped or discolored medium.
  • The prepared broth is clear and purple.
  • The pH for double-concentrated can differ from single-concentrated DEV Lactose Peptone broth.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service