Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.10676

Millipore

Tryptose broth

for the enrichment and cultivation of fastidious pathogenic microorganisms

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

Quality Level

form

medium granules (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

7.1-7.5 (25 °C, 26 g/L in H2O, after autoclaving)

solubility

26 g/L

application(s)

clinical
microbiology

storage temp.

15-25°C

suitability

Listeria spp.
Pneumococcus spp.
Streptococcus spp.

General description

for the enrichment and cultivation of fastidious pathogenic microorganisms

Analysis Note

Appearance (clearness)clearAppearance (colour)yellowish-brownpH-value (25 °C)7.1 - 7.5Growth (Streptococcus pyogenes ATCC 12344)good to very goodGrowth (Streptococcus pneumoniae ATCC 6301)good to very goodGrowth (Pasteurella multocida ATCC 43137)fair to goodGrowth (Listeria monocytogenes ATCC 19118)good to very goodGrowth (Shigella flexneri ATCC 12022 (WDCM 00126))good to very goodGrowth (Escherichia coli ATCC 25922 (WDCM 00013))good to very goodGrowth (Staphylococcus aureus ATCC 25923 (WDCM 00034))good to very goodIncubation: 24 hrs.; 35 °C; aerobic.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service