Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.10658

Millipore

M broth

suitable for microbiology

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$190.00

$190.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
500 G
$190.00

About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

$190.00


Please contact Customer Service for Availability

Request more information

form

powder (dehydrated (DCM))

Quality Level

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Salmonella spp.

General description

for microbiology

Analysis Note

Appearance (clearness): clear
Appearance (colour): yellowish-brown
pH-value (25 °C): 6.8 - 7.2
Growth (Salmonella choleraesuis ATCC 13312): good to very good
Growth (Salmonella typhimurium ATCC 14028 (WDCM 00031)): good to very good
Growth (Salmonella enteritidis ATCC 13076 (WDCM 00030)): good to very good
Incubation: 24 hrs.; 35 °C; aerobic.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service